missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-nitrotoluene 98.0+%, TCI America™

$122.84 - $361.59

Chemical Identifiers

CAS 41085-43-2
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00134555
InChI Key GCAAVRIWNMTOKB-UHFFFAOYSA-N
Synonym 2-bromo-3-nitrotoluene, 2-bromo-3-methylnitrobenzen, 2-bromo-3-methylnitrobenzene, benzene, 2-bromo-1-methyl-3-nitro, 2-bromo-1-methyl-3-nitro-benzene, 2-brom-1-methyl-3-nitrobenzol, pubchem4531, acmc-1ajt4, ksc493s8t
PubChem CID 231828
IUPAC Name 2-bromo-1-methyl-3-nitrobenzene
SMILES CC1=CC=CC(=C1Br)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37121G
View Documents
TCI America
B37121G
1 g
Each for $122.84
 
B37125G
View Documents
TCI America
B37125G
5 g
Each for $361.59
 
Description

Description

Specifications

Chemical Identifiers

41085-43-2
216.03
GCAAVRIWNMTOKB-UHFFFAOYSA-N
231828
CC1=CC=CC(=C1Br)[N+]([O-])=O
C7H6BrNO2
MFCD00134555
2-bromo-3-nitrotoluene, 2-bromo-3-methylnitrobenzen, 2-bromo-3-methylnitrobenzene, benzene, 2-bromo-1-methyl-3-nitro, 2-bromo-1-methyl-3-nitro-benzene, 2-brom-1-methyl-3-nitrobenzol, pubchem4531, acmc-1ajt4, ksc493s8t
2-bromo-1-methyl-3-nitrobenzene

Specifications

41085-43-2
Yellow
C7H6BrNO2
1 g
GCAAVRIWNMTOKB-UHFFFAOYSA-N
2-bromo-1-methyl-3-nitrobenzene
231828
≥98.0% (GC)
2-Bromo-3-nitrotoluene
40°C
136°C
MFCD00134555
2-bromo-3-nitrotoluene, 2-bromo-3-methylnitrobenzen, 2-bromo-3-methylnitrobenzene, benzene, 2-bromo-1-methyl-3-nitro, 2-bromo-1-methyl-3-nitro-benzene, 2-brom-1-methyl-3-nitrobenzol, pubchem4531, acmc-1ajt4, ksc493s8t
CC1=CC=CC(=C1Br)[N+]([O-])=O
216.03
216.03
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'