missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3'-(trifluoromethyl)acetophenone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3152606

 View more versions of this product

Catalog No. AAH3152606


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-3'-(trifluoromethyl)acetophenone
20°C to 23°C
C9H6BrF3O
5 g
TZIYNLSEBAYCBZ-UHFFFAOYSA-N
2-bromo-1-[3-(trifluoromethyl)phenyl]ethanone
2778386
98%
2003-10-3
69°C to 72°C
MFCD03094283
2-bromo-1-3-trifluoromethyl phenyl ethanone, 3-trifluoromethyl phenacyl bromide, 2-bromo-3'-trifluoromethylacetophenone, 2-bromo-1-3-trifluoromethyl phenyl ethan-1-one, 2-bromo-3'-trifluoromethyl acetophenone, 2-bromo-1-3-trifluoromethyl-phenyl-ethanone, 2-bromo-1-3-trifluoromethyl phenyl-1-ethanone, ethanone, 2-bromo-1-3-trifluoromethyl phenyl, 3-trifluoromethylphenacylbromide
C1=CC(=CC(=C1)C(F)(F)F)C(=O)CBr
267.05
267.04

Chemical Identifiers

2003-10-3
267.05
TZIYNLSEBAYCBZ-UHFFFAOYSA-N
C9H6BrF3O
MFCD03094283
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only