missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4-chloro-1-iodobenzene, 98%, Thermo Scientific Chemicals

$95.33 - $317.75

Chemical Identifiers

CAS 31928-44-6
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.348
MDL Number MFCD00672944
InChI Key CXHXFDQEFKFYQJ-UHFFFAOYSA-N
Synonym 2-bromo-4-chloroiodobenzene, benzene, 2-bromo-4-chloro-1-iodo, acmc-209hpo, ksc494q6f, 1-iodo-2-bromo-4-chlorobenzene, 2-bromo-1-iodo-4-chlorobenzene
PubChem CID 13741326
IUPAC Name 2-bromo-4-chloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Br)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2540906
View Documents
Thermo Scientific Chemicals
B2540906
5 g
Each for $95.33
 
AAB2540914
View Documents
Thermo Scientific Chemicals
B2540914
25 g
Each for $317.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

31928-44-6
317.348
CXHXFDQEFKFYQJ-UHFFFAOYSA-N
13741326
C1=CC(=C(C=C1Cl)Br)I
C6H3BrClI
MFCD00672944
2-bromo-4-chloroiodobenzene, benzene, 2-bromo-4-chloro-1-iodo, acmc-209hpo, ksc494q6f, 1-iodo-2-bromo-4-chlorobenzene, 2-bromo-1-iodo-4-chlorobenzene
2-bromo-4-chloro-1-iodobenzene

Specifications

31928-44-6
MFCD00672944
Light sensitive
CXHXFDQEFKFYQJ-UHFFFAOYSA-N
2-bromo-4-chloro-1-iodobenzene
13741326
98%
C6H3BrClI
5 g
2-bromo-4-chloroiodobenzene, benzene, 2-bromo-4-chloro-1-iodo, acmc-209hpo, ksc494q6f, 1-iodo-2-bromo-4-chlorobenzene, 2-bromo-1-iodo-4-chlorobenzene
C1=CC(=C(C=C1Cl)Br)I
317.348
317.36
2-Bromo-4-chloro-1-iodobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only