Learn More
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline, 98%
CAS: 875664-27-0 | C7H4BrF4N | 258.014 g/mol
$397.19 - $397.19
Chemical Identifiers
| CAS | 875664-27-0 |
|---|---|
| Molecular Formula | C7H4BrF4N |
| Molecular Weight (g/mol) | 258.014 |
| MDL Number | MFCD04973756 |
| InChI Key | OTYCOVZULSPPPW-UHFFFAOYSA-N |
| Synonym | 2-bromo-4-fluoro-6-trifluoromethyl aniline, 2-amino-3-bromo-5-fluorobenzotrifluoride, buttpark 154\11-38, benzenamine,2-bromo-4-fluoro-6-trifluoromethyl, 2-bromo-4-fluoro-6-trifluoromethyl benzenamine, pubchem19869, ksc658m8n, 2-bromo-4-fluoro-6-trifluoromethyl phenylamine, 6-bromo-4-fluoro-2-trifluoromethyl phenylamine, 2-bromanyl-4-fluoranyl-6-trifluoromethyl aniline |
| PubChem CID | 7018045 |
| IUPAC Name | 2-bromo-4-fluoro-6-(trifluoromethyl)aniline |
| SMILES | C1=C(C=C(C(=C1Br)N)C(F)(F)F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2620609
|
Thermo Scientific Chemicals
H2620609 |
10 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 875664-27-0 | |
| 258.014 | |
| OTYCOVZULSPPPW-UHFFFAOYSA-N | |
| 7018045 | |
| C1=C(C=C(C(=C1Br)N)C(F)(F)F)F |
| C7H4BrF4N | |
| MFCD04973756 | |
| 2-bromo-4-fluoro-6-trifluoromethyl aniline, 2-amino-3-bromo-5-fluorobenzotrifluoride, buttpark 154\11-38, benzenamine,2-bromo-4-fluoro-6-trifluoromethyl, 2-bromo-4-fluoro-6-trifluoromethyl benzenamine, pubchem19869, ksc658m8n, 2-bromo-4-fluoro-6-trifluoromethyl phenylamine, 6-bromo-4-fluoro-2-trifluoromethyl phenylamine, 2-bromanyl-4-fluoranyl-6-trifluoromethyl aniline | |
| 2-bromo-4-fluoro-6-(trifluoromethyl)aniline |
Specifications
| 875664-27-0 | |
| 1.503 | |
| 10 g | |
| 2-bromo-4-fluoro-6-trifluoromethyl aniline, 2-amino-3-bromo-5-fluorobenzotrifluoride, buttpark 154\11-38, benzenamine,2-bromo-4-fluoro-6-trifluoromethyl, 2-bromo-4-fluoro-6-trifluoromethyl benzenamine, pubchem19869, ksc658m8n, 2-bromo-4-fluoro-6-trifluoromethyl phenylamine, 6-bromo-4-fluoro-2-trifluoromethyl phenylamine, 2-bromanyl-4-fluoranyl-6-trifluoromethyl aniline | |
| C1=C(C=C(C(=C1Br)N)C(F)(F)F)F | |
| 258.014 | |
| 258.01 | |
| 2-Bromo-4-fluoro-6-(trifluoromethyl)aniline |
| C7H4BrF4N | |
| MFCD04973756 | |
| UN2810 | |
| OTYCOVZULSPPPW-UHFFFAOYSA-N | |
| 2-bromo-4-fluoro-6-(trifluoromethyl)aniline | |
| 7018045 | |
| 98% |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only