missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-4-fluorobenzyl Alcohol 98.0+%, TCI America™
$137.70 - $366.63
Chemical Identifiers
CAS | 229027-89-8 |
---|---|
Molecular Formula | C7H6BrFO |
Molecular Weight (g/mol) | 205.03 |
MDL Number | MFCD00672925 |
InChI Key | XGAMLBPEVCLQEJ-UHFFFAOYSA-N |
Synonym | 2-bromo-4-fluorophenyl methanol, 2-bromo-4-fluorobenzyl alcohol, benzenemethanol, 2-bromo-4-fluoro, 2-bromo-4-fluorophenyl methan-1-ol, 2-bromo-4-fluorobenzenemethanol, benzenemethanol,2-bromo-4-fluoro, pubchem3222, acmc-1cqar, ksc550i1h, rarechem al bd 1088 |
PubChem CID | 2773347 |
IUPAC Name | (2-bromo-4-fluorophenyl)methanol |
SMILES | OCC1=C(Br)C=C(F)C=C1 |
Chemical Identifiers
229027-89-8 | |
205.03 | |
XGAMLBPEVCLQEJ-UHFFFAOYSA-N | |
2773347 | |
OCC1=C(Br)C=C(F)C=C1 |
C7H6BrFO | |
MFCD00672925 | |
2-bromo-4-fluorophenyl methanol, 2-bromo-4-fluorobenzyl alcohol, benzenemethanol, 2-bromo-4-fluoro, 2-bromo-4-fluorophenyl methan-1-ol, 2-bromo-4-fluorobenzenemethanol, benzenemethanol,2-bromo-4-fluoro, pubchem3222, acmc-1cqar, ksc550i1h, rarechem al bd 1088 | |
(2-bromo-4-fluorophenyl)methanol |
Specifications
229027-89-8 | |
White | |
MFCD00672925 | |
2-bromo-4-fluorophenyl methanol, 2-bromo-4-fluorobenzyl alcohol, benzenemethanol, 2-bromo-4-fluoro, 2-bromo-4-fluorophenyl methan-1-ol, 2-bromo-4-fluorobenzenemethanol, benzenemethanol,2-bromo-4-fluoro, pubchem3222, acmc-1cqar, ksc550i1h, rarechem al bd 1088 | |
OCC1=C(Br)C=C(F)C=C1 | |
205.03 | |
205.03 | |
Crystalline Powder |
70°C | |
C7H6BrFO | |
5 g | |
XGAMLBPEVCLQEJ-UHFFFAOYSA-N | |
(2-bromo-4-fluorophenyl)methanol | |
2773347 | |
≥98.0% (GC) | |
2-Bromo-4-fluorobenzyl Alcohol |
Safety and Handling
TSCA : No