missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoropyridine, 97%

CAS: 357927-50-5 | C5H3BrFN | 175.988 g/mol

$114.22 - $391.90

Chemical Identifiers

CAS 357927-50-5
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD09260909
InChI Key AIEATTRZKVGMBO-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-pyridine, pyridine, 2-bromo-4-fluoro, 4-fluoro-2-bromopyridine, pubchem3004, acmc-209ij7, pybr71, ksc493m3n, 2-bromo-4-fluoropyridine, abbypharma ap-17-5103
PubChem CID 21885884
IUPAC Name 2-bromo-4-fluoropyridine
SMILES C1=CN=C(C=C1F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6478503
View Documents
Thermo Scientific Chemicals
H6478503
1 g
Each for $114.22
Only null left
 
AAH6478506
View Documents
Thermo Scientific Chemicals
H6478506
5 g
Each for $391.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

357927-50-5
175.988
AIEATTRZKVGMBO-UHFFFAOYSA-N
21885884
C1=CN=C(C=C1F)Br
C5H3BrFN
MFCD09260909
2-bromo-4-fluoro-pyridine, pyridine, 2-bromo-4-fluoro, 4-fluoro-2-bromopyridine, pubchem3004, acmc-209ij7, pybr71, ksc493m3n, 2-bromo-4-fluoropyridine, abbypharma ap-17-5103
2-bromo-4-fluoropyridine

Specifications

357927-50-5
76°C (169°F)
1.537
1 g
2-bromo-4-fluoro-pyridine, pyridine, 2-bromo-4-fluoro, 4-fluoro-2-bromopyridine, pubchem3004, acmc-209ij7, pybr71, ksc493m3n, 2-bromo-4-fluoropyridine, abbypharma ap-17-5103
C1=CN=C(C=C1F)Br
175.988
175.99
Liquid
1.699 g/mL
C5H3BrFN
MFCD09260909
14158224
AIEATTRZKVGMBO-UHFFFAOYSA-N
2-bromo-4-fluoropyridine
21885884
97%
2-Bromo-4-fluoropyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only