missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-4-fluorothiophene 95.0+%, TCI America™
$619.38 - $2157.43
Chemical Identifiers
CAS | 32431-65-5 |
---|---|
Molecular Formula | C4H2BrFS |
Molecular Weight (g/mol) | 181.022 |
InChI Key | UWXVZGDRWPSION-UHFFFAOYSA-N |
PubChem CID | 14786520 |
IUPAC Name | 2-bromo-4-fluorothiophene |
SMILES | C1=C(SC=C1F)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B3680200MG
|
TCI America
B3680200MG |
200 mg |
Each for $619.38
|
|
|||||
B36801G
|
TCI America
B36801G |
1 g |
Each for $2,157.43
|
|
|||||
Chemical Identifiers
32431-65-5 | |
181.022 | |
14786520 | |
C1=C(SC=C1F)Br |
C4H2BrFS | |
UWXVZGDRWPSION-UHFFFAOYSA-N | |
2-bromo-4-fluorothiophene |
Specifications
32431-65-5 | |
60°C | |
200 mg | |
UWXVZGDRWPSION-UHFFFAOYSA-N | |
2-bromo-4-fluorothiophene | |
14786520 | |
≥95.0% (GC) | |
2-Bromo-4-fluorothiophene |
Yellow | |
C4H2BrFS | |
1993 | |
C1=C(SC=C1F)Br | |
181.022 | |
181.02 | |
Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator