missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

$73.67 - $272.27

Chemical Identifiers

CAS 2632-13-5
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00000201
InChI Key XQJAHBHCLXUGEP-UHFFFAOYSA-N
Synonym 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone
PubChem CID 4965
IUPAC Name 2-bromo-1-(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(=O)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151900050
SDS View Documents
Thermo Scientific Chemicals
151900050
5 g Glass bottle
Each for $73.67
 
AC151900250
SDS View Documents
Thermo Scientific Chemicals
151900250
25 g Glass bottle
Each for $272.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2632-13-5
229.07
XQJAHBHCLXUGEP-UHFFFAOYSA-N
4965
COC1=CC=C(C=C1)C(=O)CBr
C9H9BrO2
MFCD00000201
2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone
2-bromo-1-(4-methoxyphenyl)ethanone

Specifications

2632-13-5
100.0
White to Brown
97.5% min. (GC)
C9H9BrO2
MFCD00000201
08, 89
2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone
XQJAHBHCLXUGEP-UHFFFAOYSA-N
2-bromo-1-(4-methoxyphenyl)ethanone
4965
98%
2-Bromo-4'-methoxyacetophenone
97.5
69°C to 72°C
Authentic
Glass bottle
CH3OC6H4COCH2Br
5 g
05,439
Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
COC1=CC=C(C=C1)C(=O)CBr
229.07
229.07
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 220-118-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only