missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4'-methylacetophenone, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 170390250

 View more versions of this product

Catalog No. AC170390250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Bromo-4'-methylacetophenone
96.0
>110°C
96% min. (GC)
C9H9BrO
MFCD00000203
07, 309
KRVGXFREOJHJAX-UHFFFAOYSA-N
2-bromo-1-(4-methylphenyl)ethanone
69272
97%
619-41-0
100.0
Authentic
Plastic bottle
CH3C6H4COCH2Br
25g
2-bromo-4'-methylacetophenone, 2-bromo-1-p-tolyl ethanone, 4-methylphenacyl bromide, 2-bromo-1-4-methylphenyl ethanone, 2-bromo-1-p-tolylethanone, 2-bromo-4-methylacetophenone, 2-bromo-1-4-methylphenyl ethan-1-one, acetophenone, 2-bromo-4'-methyl, ethanone, 2-bromo-1-4-methylphenyl, p-methylphenacyl bromide
CC1=CC=C(C=C1)C(=O)CBr
213.07
213.07

Chemical Identifiers

619-41-0
213.07
KRVGXFREOJHJAX-UHFFFAOYSA-N
69272
CC1=CC=C(C=C1)C(=O)CBr
C9H9BrO
MFCD00000203
2-bromo-4'-methylacetophenone, 2-bromo-1-p-tolyl ethanone, 4-methylphenacyl bromide, 2-bromo-1-4-methylphenyl ethanone, 2-bromo-1-p-tolylethanone, 2-bromo-4-methylacetophenone, 2-bromo-1-4-methylphenyl ethan-1-one, acetophenone, 2-bromo-4'-methyl, ethanone, 2-bromo-1-4-methylphenyl, p-methylphenacyl bromide
2-bromo-1-(4-methylphenyl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-595-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only