Learn More
2-Bromo-4'-methylpropiophenone, 98%, Thermo Scientific Chemicals
$87.44 - $1,224.78
Chemical Identifiers
CAS | 1451-82-7 |
---|---|
Molecular Formula | C10H11BrO |
Molecular Weight (g/mol) | 227.10 |
MDL Number | MFCD11131402 |
InChI Key | OZLUPIIIHOOPNQ-UHFFFAOYNA-N |
Synonym | 2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone |
PubChem CID | 2734063 |
IUPAC Name | 2-bromo-1-(4-methylphenyl)propan-1-one |
SMILES | CC(Br)C(=O)C1=CC=C(C)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6488103
|
Thermo Scientific Chemicals
H6488103 |
1 g |
N/A
|
|
|||||
AAH6488106
|
Thermo Scientific Chemicals
H6488106 |
5 g |
N/A
|
|
|||||
AAH6488114
|
Thermo Scientific Chemicals
H6488114 |
25 g |
N/A
|
|
|||||
Description
It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.
Solubility
Soluble in chloroform and methanol.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
1451-82-7 | |
227.10 | |
OZLUPIIIHOOPNQ-UHFFFAOYNA-N | |
2734063 | |
CC(Br)C(=O)C1=CC=C(C)C=C1 |
C10H11BrO | |
MFCD11131402 | |
2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone | |
2-bromo-1-(4-methylphenyl)propan-1-one |
Specifications
1451-82-7 | |
MFCD11131402 | |
2207628 | |
Soluble in chloroform and methanol. | |
CC(Br)C(=O)C1=CC=C(C)C=C1 | |
227.10 | |
227.1 | |
Solid |
C10H11BrO | |
1 g | |
2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone | |
OZLUPIIIHOOPNQ-UHFFFAOYNA-N | |
2-bromo-1-(4-methylphenyl)propan-1-one | |
2734063 | |
98% | |
2-Bromo-4'-methylpropiophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only