Learn More
2-Bromo-4-(trifluoromethyl)phenylacetonitrile, 98%, Thermo Scientific Chemicals
$103.73 - $365.23
Chemical Identifiers
CAS | 474024-36-7 |
---|---|
Molecular Formula | C9H5BrF3N |
Molecular Weight (g/mol) | 264.05 |
MDL Number | MFCD03094308 |
InChI Key | RYUZTTSDBHOIAF-UHFFFAOYSA-N |
PubChem CID | 2778436 |
IUPAC Name | 2-[2-bromo-4-(trifluoromethyl)phenyl]acetonitrile |
SMILES | FC(F)(F)C1=CC(Br)=C(CC#N)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH5351803
|
Thermo Scientific Chemicals
H5351803 |
1 g |
Each for $103.73
|
|
||||||
AAH5351806
|
Thermo Scientific Chemicals
H5351806 |
5 g |
Each for $365.23
|
|
||||||
Description
2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
474024-36-7 | |
264.05 | |
RYUZTTSDBHOIAF-UHFFFAOYSA-N | |
2-[2-bromo-4-(trifluoromethyl)phenyl]acetonitrile |
C9H5BrF3N | |
MFCD03094308 | |
2778436 | |
FC(F)(F)C1=CC(Br)=C(CC#N)C=C1 |
Specifications
474024-36-7 | |
265°C to 266°C | |
1.502 | |
1 g | |
Slightly soluble in water. | |
FC(F)(F)C1=CC(Br)=C(CC#N)C=C1 | |
264.05 | |
264.04 | |
2-Bromo-4-(trifluoromethyl)phenylacetonitrile |
1.626 | |
C9H5BrF3N | |
MFCD03094308 | |
2-bromo-4-trifluoromethyl phenylacetonitrile, 2-2-bromo-4-trifluoromethyl phenyl acetonitrile, 2-2-bromo-4-trifluoromethyl phenyl ethanenitrile, pubchem4248, 2-bromo-4-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile,2-bromo-4-trifluoromethyl, benzeneacetonitrile, 2-bromo-4-trifluoromethyl | |
RYUZTTSDBHOIAF-UHFFFAOYSA-N | |
2-[2-bromo-4-(trifluoromethyl)phenyl]acetonitrile | |
2778436 | |
98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only