missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorobenzaldehyde, 97%

CAS: 174265-12-4 | C7H4BrClO | 219.462 g/mol

$53.32 - $566.66

Chemical Identifiers

CAS 174265-12-4
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00462870
InChI Key IIISHLMCTDMUHH-UHFFFAOYSA-N
Synonym 5-chloro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-5-chloro, pubchem19849, acmc-1bwmm, ksc174i5n, 2-bromo-5-chlorobenzaldehycle, iiishlmctdmuhh-uhfffaoysa
PubChem CID 15391279
IUPAC Name 2-bromo-5-chlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)C=O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6433006
View Documents
Thermo Scientific Chemicals
H6433006
5 g
Each for $53.32
Only null left
 
AAH6433014
View Documents
Thermo Scientific Chemicals
H6433014
25 g
Each for $146.00
Only null left
 
AAH6433022
View Documents
Thermo Scientific Chemicals
H6433022
100 g
Each for $566.66
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

174265-12-4
219.462
IIISHLMCTDMUHH-UHFFFAOYSA-N
15391279
C1=CC(=C(C=C1Cl)C=O)Br
C7H4BrClO
MFCD00462870
5-chloro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-5-chloro, pubchem19849, acmc-1bwmm, ksc174i5n, 2-bromo-5-chlorobenzaldehycle, iiishlmctdmuhh-uhfffaoysa
2-bromo-5-chlorobenzaldehyde

Specifications

174265-12-4
Yellow
MFCD00462870
10423891
IIISHLMCTDMUHH-UHFFFAOYSA-N
2-bromo-5-chlorobenzaldehyde
15391279
97%
2-Bromo-5-chlorobenzaldehyde
64°C to 68°C (literature)
C7H4BrClO
5 g
5-chloro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-5-chloro, pubchem19849, acmc-1bwmm, ksc174i5n, 2-bromo-5-chlorobenzaldehycle, iiishlmctdmuhh-uhfffaoysa
C1=CC(=C(C=C1Cl)C=O)Br
219.462
219.46
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only