Learn More
2-Bromo-5-chloropyridine, 98%
CAS: 40473-01-6 | C5H3BrClN | 192.44 g/mol
$172.43 - $172.43
Chemical Identifiers
| CAS | 40473-01-6 |
|---|---|
| Molecular Formula | C5H3BrClN |
| Molecular Weight (g/mol) | 192.44 |
| MDL Number | MFCD00234006 |
| InChI Key | BZUUVQCSPHPUQA-UHFFFAOYSA-N |
| Synonym | 5-chloro-2-bromopyridine, 2-bromo-5-chloro-pyridine, 2-brom-5-chlorpyridin, pyridine, 2-bromo-5-chloro, pubchem1169, acmc-209jeb, 2-bromo-5-chloro pyridine, ksc238i0d, tpc-py102, 2-bromanyl-5-chloranyl-pyridine |
| PubChem CID | 817098 |
| IUPAC Name | 2-bromo-5-chloropyridine |
| SMILES | C1=CC(=NC=C1Cl)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC378980010
|
Thermo Scientific Chemicals
378980010 |
1 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 40473-01-6 | |
| 192.44 | |
| BZUUVQCSPHPUQA-UHFFFAOYSA-N | |
| 817098 | |
| C1=CC(=NC=C1Cl)Br |
| C5H3BrClN | |
| MFCD00234006 | |
| 5-chloro-2-bromopyridine, 2-bromo-5-chloro-pyridine, 2-brom-5-chlorpyridin, pyridine, 2-bromo-5-chloro, pubchem1169, acmc-209jeb, 2-bromo-5-chloro pyridine, ksc238i0d, tpc-py102, 2-bromanyl-5-chloranyl-pyridine | |
| 2-bromo-5-chloropyridine |
Specifications
| 40473-01-6 | |
| 100.0 | |
| Beige to Brown-Yellow | |
| 97.5% min. (GC) | |
| C5H3BrClN | |
| 1 g | |
| BZUUVQCSPHPUQA-UHFFFAOYSA-N | |
| 2-bromo-5-chloropyridine | |
| 817098 | |
| 98% | |
| 2-Bromo-5-chloropyridine |
| 97.5 | |
| 65°C to 69°C | |
| Authentic | |
| Glass bottle | |
| MFCD00234006 | |
| 5-chloro-2-bromopyridine, 2-bromo-5-chloro-pyridine, 2-brom-5-chlorpyridin, pyridine, 2-bromo-5-chloro, pubchem1169, acmc-209jeb, 2-bromo-5-chloro pyridine, ksc238i0d, tpc-py102, 2-bromanyl-5-chloranyl-pyridine | |
| C1=CC(=NC=C1Cl)Br | |
| 192.44 | |
| 192.44 | |
| Powder |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only