Learn More
2-Bromo-5-fluorobenzyl alcohol, 97%, Thermo Scientific Chemicals
$383.49 - $383.49
Chemical Identifiers
CAS | 202865-66-5 |
---|---|
Molecular Formula | C7H6BrFO |
Molecular Weight (g/mol) | 205.03 |
MDL Number | MFCD00142876 |
InChI Key | HXGZPMHPSBJMKB-UHFFFAOYSA-N |
Synonym | 2-bromo-5-fluorobenzyl alcohol, 2-bromo-5-fluorophenyl methanol, 2-bromo-5-fluorobenzylalcohol, benzenemethanol, 2-bromo-5-fluoro, 2-bromo-5-fluorophenyl methan-1-ol, buttpark 87\01-69, 2-bromo-5-fluorbenzyl alcohol, pubchem1963, acmc-1cs63, bromo-5-fluorobenzyl alcohol |
PubChem CID | 2773349 |
IUPAC Name | (2-bromo-5-fluorophenyl)methanol |
SMILES | OCC1=C(Br)C=CC(F)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6159606
|
Thermo Scientific Chemicals
H6159606 |
5 g |
N/A
|
|
||||||
Description
2-Bromo-5-fluorobenzyl alcohol is used as intermediates in pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-5-fluorobenzyl alcohol is used as intermediates in pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
Chemical Identifiers
202865-66-5 | |
205.03 | |
HXGZPMHPSBJMKB-UHFFFAOYSA-N | |
2773349 | |
OCC1=C(Br)C=CC(F)=C1 |
C7H6BrFO | |
MFCD00142876 | |
2-bromo-5-fluorobenzyl alcohol, 2-bromo-5-fluorophenyl methanol, 2-bromo-5-fluorobenzylalcohol, benzenemethanol, 2-bromo-5-fluoro, 2-bromo-5-fluorophenyl methan-1-ol, buttpark 87\01-69, 2-bromo-5-fluorbenzyl alcohol, pubchem1963, acmc-1cs63, bromo-5-fluorobenzyl alcohol | |
(2-bromo-5-fluorophenyl)methanol |
Specifications
202865-66-5 | |
Yellow | |
MFCD00142876 | |
2-bromo-5-fluorobenzyl alcohol, 2-bromo-5-fluorophenyl methanol, 2-bromo-5-fluorobenzylalcohol, benzenemethanol, 2-bromo-5-fluoro, 2-bromo-5-fluorophenyl methan-1-ol, buttpark 87\01-69, 2-bromo-5-fluorbenzyl alcohol, pubchem1963, acmc-1cs63, bromo-5-fluorobenzyl alcohol | |
HXGZPMHPSBJMKB-UHFFFAOYSA-N | |
(2-bromo-5-fluorophenyl)methanol | |
2773349 | |
97% | |
2-Bromo-5-fluorobenzyl alcohol |
91°C to 94°C | |
C7H6BrFO | |
5 g | |
Slightly soluble in water. | |
OCC1=C(Br)C=CC(F)=C1 | |
205.03 | |
205.02 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only