Learn More
2-Bromo-5-fluorophenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1506414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-5-fluorophenol | |
1.717 | |
87°C (188°F) | |
1.554 | |
25 g | |
phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843 | |
C1=CC(=C(C=C1F)O)Br | |
190.999 | |
191 |
147460-41-1 | |
184°C to 186°C | |
C6H4BrFO | |
MFCD00040939 | |
7915214 | |
HUVAOAVBKOVPBZ-UHFFFAOYSA-N | |
2-bromo-5-fluorophenol | |
2724600 | |
97% |
Chemical Identifiers
147460-41-1 | |
190.999 | |
HUVAOAVBKOVPBZ-UHFFFAOYSA-N | |
2724600 | |
C1=CC(=C(C=C1F)O)Br |
C6H4BrFO | |
MFCD00040939 | |
phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843 | |
2-bromo-5-fluorophenol |
Safety and Handling
GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only