missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-5-hydroxypyridine, 95%, Thermo Scientific Chemicals

$180.42 - $405.41

Chemical Identifiers

CAS 55717-45-8
Molecular Formula C5H6BNO
Molecular Weight (g/mol) 106.92
MDL Number MFCD04114184
InChI Key ADHMWMAEKRZWPB-UHFFFAOYSA-N
Synonym 2-bromo-5-hydroxypyridine, 6-bromo-3-pyridinol, 6-bromo-3-hydroxypyridine, 3-pyridinol, 6-bromo, 5-hydroxy-2-bromopyridine, pubchem5967, zlchem 1240, 2-bromopyridine-5-ol, acmc-209lor, 3-hydroxy-6-bromopyridine
PubChem CID 2762865
IUPAC Name 6-bromopyridin-3-ol
SMILES BC1=CC=C(O)C=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2722803
View Documents
Thermo Scientific Chemicals
H2722803
1 g
Each for $180.42
 
AAH2722806
View Documents
Thermo Scientific Chemicals
H2722806
5 g
Each for $405.41
 
Description

Description

It is used as an active pharmaceutical intermediate and in the coupling reactions with halo aromatic amines and alcohols for a practical synthetic route to 2-substituted amino phenyl and hydroxyphenyl pyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and in the coupling reactions with halo aromatic amines and alcohols for a practical synthetic route to 2-substituted amino phenyl and hydroxyphenyl pyridines.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

55717-45-8
106.92
ADHMWMAEKRZWPB-UHFFFAOYSA-N
2762865
BC1=CC=C(O)C=N1
C5H6BNO
MFCD04114184
2-bromo-5-hydroxypyridine, 6-bromo-3-pyridinol, 6-bromo-3-hydroxypyridine, 3-pyridinol, 6-bromo, 5-hydroxy-2-bromopyridine, pubchem5967, zlchem 1240, 2-bromopyridine-5-ol, acmc-209lor, 3-hydroxy-6-bromopyridine
6-bromopyridin-3-ol

Specifications

55717-45-8
C5H6BNO
1 g
Slightly soluble in water.
BC1=CC=C(O)C=N1
106.92
174
2-Bromo-5-hydroxypyridine
133°C to 136°C
MFCD04114184
2-bromo-5-hydroxypyridine, 6-bromo-3-pyridinol, 6-bromo-3-hydroxypyridine, 3-pyridinol, 6-bromo, 5-hydroxy-2-bromopyridine, pubchem5967, zlchem 1240, 2-bromopyridine-5-ol, acmc-209lor, 3-hydroxy-6-bromopyridine
ADHMWMAEKRZWPB-UHFFFAOYSA-N
6-bromopyridin-3-ol
2762865
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only