missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitroanisole 98.0+%, TCI America™

$143.54 - $570.40

Chemical Identifiers

CAS 77337-82-7
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD00041250
InChI Key NTKADLOYTKVXQN-UHFFFAOYSA-N
Synonym 2-bromo-5-nitroanisole, 4-bromo-3-methoxynitrobenzene, 2-bromo-5-nitroanisol, benzene, 1-bromo-2-methoxy-4-nitro, 1-bromo-2-methoxy-4-nitro-benzene, zlchem 673, pubchem2536, 2-bromo 5-nitro anisole, 2-bromo-5-nitro-anisole, 2-bromo-5-nitro anisole
PubChem CID 101293
IUPAC Name 1-bromo-2-methoxy-4-nitrobenzene
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B34185G
View Documents
TCI America
B34185G
5 g
Each for $143.54
Only null left
 
B341825G
View Documents
TCI America
B341825G
25 g
Each for $570.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

77337-82-7
232.033
NTKADLOYTKVXQN-UHFFFAOYSA-N
101293
COC1=C(C=CC(=C1)[N+](=O)[O-])Br
C7H6BrNO3
MFCD00041250
2-bromo-5-nitroanisole, 4-bromo-3-methoxynitrobenzene, 2-bromo-5-nitroanisol, benzene, 1-bromo-2-methoxy-4-nitro, 1-bromo-2-methoxy-4-nitro-benzene, zlchem 673, pubchem2536, 2-bromo 5-nitro anisole, 2-bromo-5-nitro-anisole, 2-bromo-5-nitro anisole
1-bromo-2-methoxy-4-nitrobenzene

Specifications

77337-82-7
C7H6BrNO3
5 g
NTKADLOYTKVXQN-UHFFFAOYSA-N
1-bromo-2-methoxy-4-nitrobenzene
101293
≥98.0% (GC)
2-Bromo-5-nitroanisole
104°C
MFCD00041250
2-bromo-5-nitroanisole, 4-bromo-3-methoxynitrobenzene, 2-bromo-5-nitroanisol, benzene, 1-bromo-2-methoxy-4-nitro, 1-bromo-2-methoxy-4-nitro-benzene, zlchem 673, pubchem2536, 2-bromo 5-nitro anisole, 2-bromo-5-nitro-anisole, 2-bromo-5-nitro anisole
COC1=C(C=CC(=C1)[N+](=O)[O-])Br
232.033
232.03
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'