Learn More
2-Bromo-5-nitrobenzoic acid, Thermo Scientific™
Chemical Identifiers
| CAS | 943-14-6 |
|---|---|
| Molecular Formula | C7H4BrNO4 |
| Molecular Weight (g/mol) | 246.02 |
| MDL Number | MFCD00134558 |
| InChI Key | UVFWYVCDRKRAJH-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzic acid, benzoic acid, 2-bromo-5-nitro, 2-bromo-5-nitrobenzoicacid, 2-bromo-5-nitro benzoic acid, pubchem3821, acmc-209rrc, ksc494m1r, paragos 440009, timtec-bb sbb003179 |
| PubChem CID | 243025 |
| SMILES | C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC342290050
|
Thermo Scientific Chemicals
342290050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 943-14-6 | |
| 246.02 | |
| UVFWYVCDRKRAJH-UHFFFAOYSA-N | |
| 243025 |
| C7H4BrNO4 | |
| MFCD00134558 | |
| 2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzic acid, benzoic acid, 2-bromo-5-nitro, 2-bromo-5-nitrobenzoicacid, 2-bromo-5-nitro benzoic acid, pubchem3821, acmc-209rrc, ksc494m1r, paragos 440009, timtec-bb sbb003179 | |
| C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br |
Specifications
| 943-14-6 | |
| Glass bottle | |
| MFCD00134558 | |
| 2-bromo-5-nitrobenzoic acid, 2-bromo-5-nitrobenzic acid, benzoic acid, 2-bromo-5-nitro, 2-bromo-5-nitrobenzoicacid, 2-bromo-5-nitro benzoic acid, pubchem3821, acmc-209rrc, ksc494m1r, paragos 440009, timtec-bb sbb003179 | |
| UVFWYVCDRKRAJH-UHFFFAOYSA-N | |
| 246.02 | |
| 246.02 | |
| 2-Bromo-5-nitrobenzoic acid |
| Authentic | |
| C7H4BrNO4 | |
| 5g | |
| Solubility in water: insoluble | |
| C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Br | |
| 243025 | |
| ≥96.0% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA
RUO â Research Use Only