missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrofuran 98.0+%, TCI America™

$244.19 - $809.00

Chemical Identifiers

CAS 823-73-4
Molecular Formula C4H2BrNO3
Molecular Weight (g/mol) 191.97
MDL Number MFCD00142879
InChI Key QGTDMAKRJHGXOF-UHFFFAOYSA-N
PubChem CID 233582
IUPAC Name 2-bromo-5-nitrofuran
SMILES [O-][N+](=O)C1=CC=C(Br)O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14701G
View Documents
TCI America
B14701G
1 g N/A
Only null left
 
B14705G
View Documents
TCI America
B14705G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

823-73-4
191.97
QGTDMAKRJHGXOF-UHFFFAOYSA-N
2-bromo-5-nitrofuran
C4H2BrNO3
MFCD00142879
233582
[O-][N+](=O)C1=CC=C(Br)O1

Specifications

823-73-4
Yellow
C4H2BrNO3
1 g
[O-][N+](=O)C1=CC=C(Br)O1
191.97
191.97
Crystalline Powder
47°C
118°C
MFCD00142879
QGTDMAKRJHGXOF-UHFFFAOYSA-N
2-bromo-5-nitrofuran
233582
≥98.0% (GC)
2-Bromo-5-nitrofuran
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'