missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-6-methoxypyridine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 397670010

 View more versions of this product

Catalog No. AC397670010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Bromo-6-methoxypyridine
96.0
1.5300g/mL
104°C
96% min. (GC)
C6H6BrNO
1mL
2-bromo-6-methoxy-pyridine, pyridine, 2-bromo-6-methoxy, 6-bromo-2-methoxypyridine, 2-methoxy-6-bromo pyridine, buttpark 82\06-06, 2-brom-6-methoxypyridin, zlchem 781, pubchem9206, acmc-209jec, 2-bromo-6-methoxy-pyridin
COC1=CC=CC(Br)=N1
188.02
188.02
40473-07-2
100.0
206°C
Authentic
Glass bottle
MFCD00088345
1.53
KMODISUYWZPVGV-UHFFFAOYSA-N
2-bromo-6-methoxypyridine
256810
97%

Chemical Identifiers

40473-07-2
188.02
KMODISUYWZPVGV-UHFFFAOYSA-N
256810
COC1=CC=CC(Br)=N1
C6H6BrNO
MFCD00088345
2-bromo-6-methoxy-pyridine, pyridine, 2-bromo-6-methoxy, 6-bromo-2-methoxypyridine, 2-methoxy-6-bromo pyridine, buttpark 82\06-06, 2-brom-6-methoxypyridin, zlchem 781, pubchem9206, acmc-209jec, 2-bromo-6-methoxy-pyridin
2-bromo-6-methoxypyridine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only