missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-L-phenylalanine, 95%, Thermo Scientific Chemicals
$155.96 - $606.29
Chemical Identifiers
CAS | 42538-40-9 |
---|---|
Molecular Formula | C9H10BrNO2 |
Molecular Weight (g/mol) | 244.09 |
MDL Number | MFCD03092958 |
InChI Key | JFVLNTLXEZDFHW-QMMMGPOBSA-N |
Synonym | l-2-bromophenylalanine, 2-bromo-l-phenylalanine, s-2-amino-3-2-bromophenyl propanoic acid, 2-bromophenylalanine, 2s-2-amino-3-2-bromophenyl propanoic acid, h-phe 2-br-oh, o-bromo-l-phenylalanine, l-2-br-phe-oh, s-2-bromophenylalanine, l-2-bromophe |
PubChem CID | 193338 |
IUPAC Name | (2S)-2-amino-3-(2-bromophenyl)propanoic acid |
SMILES | N[C@@H](CC1=CC=CC=C1Br)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6314103
|
Thermo Scientific Chemicals
H6314103 |
1 g |
N/A
|
|
||||||
AAH6314106
|
Thermo Scientific Chemicals
H6314106 |
5 g |
N/A
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
42538-40-9 | |
244.09 | |
JFVLNTLXEZDFHW-QMMMGPOBSA-N | |
193338 | |
N[C@@H](CC1=CC=CC=C1Br)C(O)=O |
C9H10BrNO2 | |
MFCD03092958 | |
l-2-bromophenylalanine, 2-bromo-l-phenylalanine, s-2-amino-3-2-bromophenyl propanoic acid, 2-bromophenylalanine, 2s-2-amino-3-2-bromophenyl propanoic acid, h-phe 2-br-oh, o-bromo-l-phenylalanine, l-2-br-phe-oh, s-2-bromophenylalanine, l-2-bromophe | |
(2S)-2-amino-3-(2-bromophenyl)propanoic acid |
Specifications
42538-40-9 | |
C9H10BrNO2 | |
l-2-bromophenylalanine, 2-bromo-l-phenylalanine, s-2-amino-3-2-bromophenyl propanoic acid, 2-bromophenylalanine, 2s-2-amino-3-2-bromophenyl propanoic acid, h-phe 2-br-oh, o-bromo-l-phenylalanine, l-2-br-phe-oh, s-2-bromophenylalanine, l-2-bromophe | |
N[C@@H](CC1=CC=CC=C1Br)C(O)=O | |
1 g | |
193338 | |
95% | |
2-Bromo-L-phenylalanine |
White | |
MFCD03092958 | |
JFVLNTLXEZDFHW-QMMMGPOBSA-N | |
(2S)-2-amino-3-(2-bromophenyl)propanoic acid | |
244.09 | |
244.09 | |
Powder |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only