Learn More
2-Bromo-m-xylene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 152030250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-m-xylene | |
576-22-7 | |
97.5 | |
1.3800g/mL | |
25 g | |
Glass bottle | |
1.5540 to 1.556 | |
1.38 | |
(CH3)2C6H3Br | |
05, 374 | |
MYMYVYZLMUEVED-UHFFFAOYSA-N | |
2-bromo-1,3-dimethylbenzene | |
68471 | |
98% |
-10°C | |
Colorless to Yellow | |
100.0 | |
206°C | |
Authentic | |
Liquid | |
98% | |
C8H9Br | |
MFCD00000075 | |
2,6-dimethylbromobenzene, 2-bromo-m-xylene, 1-bromo-2,6-dimethylbenzene, benzene, 2-bromo-1,3-dimethyl, 2,6-dimethyl bromobenzene, m-xylene, 2-bromo, 2,6-dimethylphenyl bromide, 2-bromo-meta-xylene, 2-bromo-1,3-dimethyl-benzene, pubchem3206 | |
CC1=C(C(=CC=C1)C)Br | |
185.06 | |
185.06 |
Chemical Identifiers
576-22-7 | |
185.06 | |
MYMYVYZLMUEVED-UHFFFAOYSA-N | |
68471 | |
CC1=C(C(=CC=C1)C)Br |
C8H9Br | |
MFCD00000075 | |
2,6-dimethylbromobenzene, 2-bromo-m-xylene, 1-bromo-2,6-dimethylbenzene, benzene, 2-bromo-1,3-dimethyl, 2,6-dimethyl bromobenzene, m-xylene, 2-bromo, 2,6-dimethylphenyl bromide, 2-bromo-meta-xylene, 2-bromo-1,3-dimethyl-benzene, pubchem3206 | |
2-bromo-1,3-dimethylbenzene |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
EINECSNumber : 209-397-7
RUO – Research Use Only