missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-N,N-diethylethylamine hydrobromide, 98%, Thermo Scientific™

Chemical Identifiers

CAS 1069-72-3
Molecular Formula C6H15Br2N
Molecular Weight (g/mol) 261.00
MDL Number MFCD00040376
InChI Key HLMHCDKXKXBKQK-UHFFFAOYSA-N
Synonym 2-diethylamino ethyl bromide hydrobromide, 2-bromo-n,n-diethylethylamine hydrobromide, 2-bromoethyldiethylammonium bromide, 2-bromoethyl diethylamine hydrobromide, 2-bromo-n,n-diethylethanaminium bromide, 2-bromotriethylamine hydrobromide, 2-bromoethyldiethylamine hydrobromide, diethylaminoethyl bromide hydrobromide, 2-bromo-n,n-diethylethanamine hydrobromide, ethanamine, 2-bromo-n,n-diethyl-, hydrobromide
PubChem CID 70607
IUPAC Name 2-bromo-N,N-diethylethanamine;hydrobromide
SMILES [H+].[Br-].CCN(CC)CCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426860250
SDS View Documents
Thermo Scientific Chemicals
426860250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC426861000
SDS View Documents
Thermo Scientific Chemicals
426861000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1069-72-3
261.00
HLMHCDKXKXBKQK-UHFFFAOYSA-N
70607
[H+].[Br-].CCN(CC)CCBr
C6H15Br2N
MFCD00040376
2-diethylamino ethyl bromide hydrobromide, 2-bromo-n,n-diethylethylamine hydrobromide, 2-bromoethyldiethylammonium bromide, 2-bromoethyl diethylamine hydrobromide, 2-bromo-n,n-diethylethanaminium bromide, 2-bromotriethylamine hydrobromide, 2-bromoethyldiethylamine hydrobromide, diethylaminoethyl bromide hydrobromide, 2-bromo-n,n-diethylethanamine hydrobromide, ethanamine, 2-bromo-n,n-diethyl-, hydrobromide
2-bromo-N,N-diethylethanamine;hydrobromide

Specifications

1069-72-3
100.0
97.5% min. (Argentometry)
C6H15Br2N
MFCD00040376
2-diethylamino ethyl bromide hydrobromide, 2-bromo-n,n-diethylethylamine hydrobromide, 2-bromoethyldiethylammonium bromide, 2-bromoethyl diethylamine hydrobromide, 2-bromo-n,n-diethylethanaminium bromide, 2-bromotriethylamine hydrobromide, 2-bromoethyldiethylamine hydrobromide, diethylaminoethyl bromide hydrobromide, 2-bromo-n,n-diethylethanamine hydrobromide, ethanamine, 2-bromo-n,n-diethyl-, hydrobromide
[H+].[Br-].CCN(CC)CCBr
261.00
260.99
2-Bromo-N, N-diethylethylamine hydrobromide
97.5
Authentic
Glass bottle
(CH3CH2)2N(CH2)2Br·HBr
25g
HLMHCDKXKXBKQK-UHFFFAOYSA-N
2-bromo-N,N-diethylethanamine;hydrobromide
70607
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-962-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only