missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoacetamide, 98%, Thermo Scientific Chemicals

 

$85.88 - $275.29

Chemical Identifiers

CAS 683-57-8
Molecular Formula C2H4BrNO
Molecular Weight (g/mol) 137.964
MDL Number MFCD00008025
InChI Key JUIKUQOUMZUFQT-UHFFFAOYSA-N
Synonym bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
PubChem CID 69632
IUPAC Name 2-bromoacetamide
SMILES C(C(=O)N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1192606
View Documents
Thermo Scientific Chemicals
A1192606
5 g
Each for $85.88
 
AAA1192614
View Documents
Thermo Scientific Chemicals
A1192614
25 g
Each for $275.29
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
Specifications

Chemical Identifiers

683-57-8
137.964
JUIKUQOUMZUFQT-UHFFFAOYSA-N
69632
C(C(=O)N)Br
C2H4BrNO
MFCD00008025
bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
2-bromoacetamide

Specifications

683-57-8
C2H4BrNO
MFCD00008025
UN2923
bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
C(C(=O)N)Br
137.964
137.97
2-Bromoacetamide
87°C to 90°C
BrCH2CONH2
5 g
1739073
JUIKUQOUMZUFQT-UHFFFAOYSA-N
2-bromoacetamide
69632
98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 000-000-0

RTECSNumber : AB4587000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only