missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Bromoallyl)trimethylsilane 90.0+%, TCI America™

$481.90 - $1668.87

Chemical Identifiers

CAS 81790-10-5
Molecular Formula C6H13BrSi
Molecular Weight (g/mol) 193.159
MDL Number MFCD00043054
InChI Key LUPQCAARZVEFMT-UHFFFAOYSA-N
PubChem CID 4173130
IUPAC Name 2-bromoprop-2-enyl(trimethyl)silane
SMILES C[Si](C)(C)CC(=C)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16951G
View Documents
TCI America
B16951G
1 g
Each for $481.90
 
B16955G
View Documents
TCI America
B16955G
5 g
Each for $1,668.87
 
Description

Description

Specifications

Chemical Identifiers

81790-10-5
193.159
LUPQCAARZVEFMT-UHFFFAOYSA-N
2-bromoprop-2-enyl(trimethyl)silane
C6H13BrSi
MFCD00043054
4173130
C[Si](C)(C)CC(=C)Br

Specifications

81790-10-5
C6H13BrSi
1 g
LUPQCAARZVEFMT-UHFFFAOYSA-N
2-bromoprop-2-enyl(trimethyl)silane
4173130
≥90.0% (GC)
(2-Bromoallyl)trimethylsilane
Yellow
MFCD00043054
1993
C[Si](C)(C)CC(=C)Br
193.159
193.16
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'