Learn More
2-Bromobenzyl alcohol, 98%, Thermo Scientific Chemicals
$85.50 - $85.50
Chemical Identifiers
CAS | 18982-54-2 |
---|---|
Molecular Formula | C7H7BrO |
Molecular Weight (g/mol) | 187.04 |
MDL Number | MFCD00004600 |
InChI Key | IOWGHQGLUMEZKG-UHFFFAOYSA-N |
Synonym | 2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746 |
PubChem CID | 72850 |
IUPAC Name | (2-bromophenyl)methanol |
SMILES | OCC1=CC=CC=C1Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC180100100
|
Thermo Scientific Chemicals
180100100 |
10 g | Glass bottle |
Each for $85.50
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18982-54-2 | |
187.04 | |
IOWGHQGLUMEZKG-UHFFFAOYSA-N | |
72850 | |
OCC1=CC=CC=C1Br |
C7H7BrO | |
MFCD00004600 | |
2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746 | |
(2-bromophenyl)methanol |
Specifications
18982-54-2 | |
100.0 | |
98% | |
C7H7BrO | |
MFCD00004600 | |
06, 445 | |
Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents | |
OCC1=CC=CC=C1Br | |
187.04 | |
187.04 | |
2-Bromobenzyl alcohol, 98% |
97.5 | |
78.0°C to 82.0°C | |
Glass bottle | |
BrC6H4CH2OH | |
10 g | |
2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746 | |
IOWGHQGLUMEZKG-UHFFFAOYSA-N | |
(2-bromophenyl)methanol | |
72850 | |
98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Do not eat,drink or smoke when using this product.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
GHS Signal Word: Warning
EINECSNumber : 242-719-4
TSCA : TSCA
RUO – Research Use Only