missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromobutane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A150690B

 View more versions of this product

Catalog No. AAA150690B


Explore available promotions

Description

Description

2-Bromobutane is utilized in the synthesis of non-nucleosides. It is used as a solvent as well as an intermediate in organic synthesis. It is an important raw material used to prepare Grignard reagent by reacting with magnesium. Further, it reacts with hydrocyanic acid to prepare sodium salt to get (+-)-sec-butyl cyanide. It is an important raw material used to prepare Grignard reagent by reacting with magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobutane is utilized in the synthesis of non-nucleosides. It is used as a solvent as well as an intermediate in organic synthesis. It is an important raw material used to prepare Grignard reagent by reacting with magnesium. Further, it reacts with hydrocyanic acid to prepare sodium salt to get (+-)-sec-butyl cyanide. It is an important raw material used to prepare Grignard reagent by reacting with magnesium.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, sodium, sodium oxides and potassium.
Specifications

Specifications

2-Bromobutane
-112°C
91°C to 92°C
98%
C4H9Br
CH3CHBrCH2CH3
1000 g
505949
sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
UPSXAPQYNGXVBF-UHFFFAOYSA-N
2-bromobutane
6554
98%
78-76-2
1.26
13°C (55°F)
Pleasant
1.4369
MFCD00000156
UN2339
14,1554
Miscible with alcohol and ether. Immiscible with water.
CCC(C)Br
137.02
137.03
Liquid

Chemical Identifiers

78-76-2
137.02
UPSXAPQYNGXVBF-UHFFFAOYSA-N
6554
CCC(C)Br
C4H9Br
MFCD00000156
sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
2-bromobutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H336
Highly flammable liquid and vapour.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOBUTANE

EINECSNumber : 201-140-7

RTECSNumber : EJ6228000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only