missing translation for 'onlineSavingsMsg'
Learn More

2-Bromocumene 97.0+%, TCI America™

$48.46 - $186.41

Chemical Identifiers

CAS 7073-94-1
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00051567
InChI Key LECYCYNAEJDSIL-UHFFFAOYSA-N
Synonym 1-bromo-2-isopropylbenzene, 1-bromo-2-1-methylethyl benzene, 2-isopropylbromobenzene, o-bromocumene, 2-bromocumene, 2-bromo cumene, cumene, o-bromo, benzene, 1-bromo-2-1-methylethyl, 1-bromo-2-propan-2-yl benzene, 1-bromo-2-isopropyl benzene
PubChem CID 23475
IUPAC Name 1-bromo-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20445G
View Documents
TCI America
B20445G
5 g
Each for $48.46
Only null left
 
B204425G
View Documents
TCI America
B204425G
25 g
Each for $186.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7073-94-1
199.091
LECYCYNAEJDSIL-UHFFFAOYSA-N
23475
CC(C)C1=CC=CC=C1Br
C9H11Br
MFCD00051567
1-bromo-2-isopropylbenzene, 1-bromo-2-1-methylethyl benzene, 2-isopropylbromobenzene, o-bromocumene, 2-bromocumene, 2-bromo cumene, cumene, o-bromo, benzene, 1-bromo-2-1-methylethyl, 1-bromo-2-propan-2-yl benzene, 1-bromo-2-isopropyl benzene
1-bromo-2-propan-2-ylbenzene

Specifications

7073-94-1
210°C
MFCD00051567
1-bromo-2-isopropylbenzene, 1-bromo-2-1-methylethyl benzene, 2-isopropylbromobenzene, o-bromocumene, 2-bromocumene, 2-bromo cumene, cumene, o-bromo, benzene, 1-bromo-2-1-methylethyl, 1-bromo-2-propan-2-yl benzene, 1-bromo-2-isopropyl benzene
CC(C)C1=CC=CC=C1Br
199.091
199.09
Liquid
Yellow
C9H11Br
5 g
LECYCYNAEJDSIL-UHFFFAOYSA-N
1-bromo-2-propan-2-ylbenzene
23475
≥97.0% (GC)
2-Bromocumene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'