Learn More
2-Bromoethyl ethyl ether, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 367405000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Bromoethyl ethyl ether | |
88.0 | |
1.3570g/mL | |
21°C | |
88% min. (GC) | |
C4H9BrO | |
BrCH2CH2OC2H5 | |
500g | |
1.357 | |
Solubility in water: slightly soluble in water. Other solubilities: miscible in ethanol,ethyl ether | |
CCOCCBr | |
153.02 | |
153.02 |
592-55-2 | |
100.0 | |
125°C to 127°C | |
Authentic | |
Glass Bottle | |
1.4435 to 1.4515 | |
MFCD00000237 | |
01,338 | |
2-bromoethyl ethyl ether, 2-ethoxyethyl bromide, 2-bromoethoxyethane, ethane, 1-bromo-2-ethoxy, 2-bromodiethyl ether, 1-ethoxy-2-bromoethane, 1-bromo-2-ethoxy-ethane, ether, 2-bromoethyl ethyl, 2-bromoethylethylether, 1-bromo-3-oxapentane | |
MMYKTRPLXXWLBC-UHFFFAOYSA-N | |
1-bromo-2-ethoxyethane | |
61141 | |
90% |
Chemical Identifiers
592-55-2 | |
153.02 | |
MMYKTRPLXXWLBC-UHFFFAOYSA-N | |
61141 | |
CCOCCBr |
C4H9BrO | |
MFCD00000237 | |
2-bromoethyl ethyl ether, 2-ethoxyethyl bromide, 2-bromoethoxyethane, ethane, 1-bromo-2-ethoxy, 2-bromodiethyl ether, 1-ethoxy-2-bromoethane, 1-bromo-2-ethoxy-ethane, ether, 2-bromoethyl ethyl, 2-bromoethylethylether, 1-bromo-3-oxapentane | |
1-bromo-2-ethoxyethane |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Danger
EINECSNumber : 209-763-6
RUO â Research Use Only