missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofluorene, 95%

CAS: 1133-80-8 | C13H9Br | 245.119 g/mol

$91.96 - $282.45

Chemical Identifiers

CAS 1133-80-8
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001115
InChI Key FXSCJZNMWILAJO-UHFFFAOYSA-N
Synonym 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
PubChem CID 14336
IUPAC Name 2-bromo-9H-fluorene
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0758906
View Documents
Thermo Scientific Chemicals
L0758906
5 g N/A
Only null left
 
AAL0758914
View Documents
Thermo Scientific Chemicals
L0758914
25 g N/A
Only null left
 
Description

Description

2-Bromofluorene acts as a nonintrusive end-capping reagent in order to control molecular weights as well as in the generation of well-defined oligomers. It is used to prepare 2-bromo-9-cyclohepta-2,4,6-trienyl-9H-fluorene and 9,9-dihexylflorene. In addition to this, it is used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromofluorene acts as a nonintrusive end-capping reagent in order to control molecular weights as well as in the generation of well-defined oligomers. It is used to prepare 2-bromo-9-cyclohepta-2,4,6-trienyl-9H-fluorene and 9,9-dihexylflorene. In addition to this, it is used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

1133-80-8
245.119
FXSCJZNMWILAJO-UHFFFAOYSA-N
14336
C1C2=CC=CC=C2C3=C1C=C(C=C3)Br
C13H9Br
MFCD00001115
2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
2-bromo-9H-fluorene

Specifications

1133-80-8
185°C (135 mmHg)
MFCD00001115
1946701
Soluble in chloroform and ethyl acetate.
C1C2=CC=CC=C2C3=C1C=C(C=C3)Br
245.119
245.12
2-Bromofluorene
110°C to 114°C
C13H9Br
5 g
2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
FXSCJZNMWILAJO-UHFFFAOYSA-N
2-bromo-9H-fluorene
14336
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-480-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only