missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals

$97.19 - $330.48

Chemical Identifiers

CAS 1072-85-1
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000282
InChI Key IPWBFGUBXWMIPR-UHFFFAOYSA-N
Synonym 2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
PubChem CID 61259
IUPAC Name 1-bromo-2-fluorobenzene
SMILES C1=CC=C(C(=C1)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC106990100
SDS View Documents
Thermo Scientific Chemicals
106990100
10 mL Glass bottle
Each for $97.19
 
AC106990500
SDS View Documents
Thermo Scientific Chemicals
106990500
50 mL Glass bottle
Each for $330.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1072-85-1
175
IPWBFGUBXWMIPR-UHFFFAOYSA-N
61259
C1=CC=C(C(=C1)F)Br
C6H4BrF
MFCD00000282
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
1-bromo-2-fluorobenzene

Specifications

1072-85-1
100.0
1.6000g/mL
43°C
99%
C6H4BrF
BrC6H4F
10 mL
1.6
Solubility in water: insoluble in water
C1=CC=C(C(=C1)F)Br
175
175
Liquid
98.5
Colorless to Yellow
155.0°C to 157.0°C
Authentic
Glass bottle
1.5327 to 1.5347
MFCD00000282
01,77
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
IPWBFGUBXWMIPR-UHFFFAOYSA-N
1-bromo-2-fluorobenzene
61259
99%
2-Bromofluorobenzene, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear pro

GHS Signal Word: Warning

EINECSNumber : 214-018-3

RTECSNumber : DA1050000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only