missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromomalonaldehyde, 97+%, Thermo Scientific Chemicals
$559.47 - $559.47
Chemical Identifiers
CAS | 2065-75-0 |
---|---|
Molecular Formula | C3H3BrO2 |
Molecular Weight (g/mol) | 150.95 |
InChI Key | SURMYNZXHKLDFO-UHFFFAOYSA-N |
Synonym | 2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde |
PubChem CID | 74945 |
IUPAC Name | 2-bromopropanedial |
SMILES | C(=O)C(C=O)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC397900250
|
Thermo Scientific Chemicals
397900250 |
25 g | Glass bottle |
Each for $559.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2065-75-0 | |
150.95 | |
2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde | |
2-bromopropanedial |
C3H3BrO2 | |
SURMYNZXHKLDFO-UHFFFAOYSA-N | |
74945 | |
C(=O)C(C=O)Br |
Specifications
2065-75-0 | |
98.5 % min. (Acid) | |
C3H3BrO2 | |
25 g | |
SURMYNZXHKLDFO-UHFFFAOYSA-N | |
2-bromopropanedial | |
74945 | |
97+% |
132.0°C to 136.0°C | |
Glass bottle | |
BrCH(CHO)2 | |
2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde | |
C(=O)C(C=O)Br | |
150.95 | |
150.95 | |
2-Bromomalonaldehyde |
Safety and Handling
GHS Signal Word: Danger
RUO – Research Use Only