missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)naphthalene, 96%

CAS: 939-26-4 | C11H9Br | 221.10 g/mol

$295.73 - $295.73

Chemical Identifiers

CAS 939-26-4
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.10
MDL Number MFCD00004123
InChI Key RUHJZSZTSCSTCC-UHFFFAOYSA-N
Synonym 2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide
PubChem CID 70320
IUPAC Name 2-(bromomethyl)naphthalene
SMILES BrCC1=CC=C2C=CC=CC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162560250
SDS View Documents
Thermo Scientific Chemicals
162560250
25 g Glass bottle
Each for $295.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

939-26-4
221.10
RUHJZSZTSCSTCC-UHFFFAOYSA-N
70320
BrCC1=CC=C2C=CC=CC2=C1
C11H9Br
MFCD00004123
2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide
2-(bromomethyl)naphthalene

Specifications

939-26-4
100.0
Yellow to Beige
>110°C
95% min. (GC)
C11H9Br
MFCD00004123
05,568
RUHJZSZTSCSTCC-UHFFFAOYSA-N
2-(bromomethyl)naphthalene
70320
96%
2-(Bromomethyl)naphthalene
95.0
52°C to 57°C
213°C (100.0 mmHg)
Authentic
Glass bottle
C10H7CH2Br
25 g
2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide
BrCC1=CC=C2C=CC=CC2=C1
221.10
221.1
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-359-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only