missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 370110050

 View more versions of this product

Catalog No. AC370110050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(Bromomethyl)pyridine hydrobromide
97.5
149.0°C to 152.0°C
Authentic
Glass bottle
BrCH2(C5H3N)
5 g
JQDNCGRNPYKRAO-UHFFFAOYSA-N
2-(bromomethyl)pyridine;hydrobromide
2734720
98%
31106-82-8
100.0
Brown to White
98%
C6H6BrN·HBr
MFCD01863544
2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide
C1=CC=NC(=C1)CBr.Br
252.94
252.94
Crystalline Powder or Crystals

Chemical Identifiers

31106-82-8
252.94
JQDNCGRNPYKRAO-UHFFFAOYSA-N
2734720
C1=CC=NC(=C1)CBr.Br
C6H6BrN·HBr
MFCD01863544
2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide
2-(bromomethyl)pyridine;hydrobromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only