missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromonicotinic Acid 98.0+%, TCI America™
$45.43 - $164.90
Chemical Identifiers
CAS | 35905-85-2 |
---|---|
Molecular Formula | C6H4BrNO2 |
Molecular Weight (g/mol) | 202.007 |
MDL Number | MFCD01646068 |
InChI Key | MMWNKXIFVYQOTK-UHFFFAOYSA-N |
Synonym | 2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f |
PubChem CID | 233262 |
IUPAC Name | 2-bromopyridine-3-carboxylic acid |
SMILES | C1=CC(=C(N=C1)Br)C(=O)O |
Chemical Identifiers
35905-85-2 | |
202.007 | |
MMWNKXIFVYQOTK-UHFFFAOYSA-N | |
233262 | |
C1=CC(=C(N=C1)Br)C(=O)O |
C6H4BrNO2 | |
MFCD01646068 | |
2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f | |
2-bromopyridine-3-carboxylic acid |
Specifications
35905-85-2 | |
C6H4BrNO2 | |
1 g | |
MMWNKXIFVYQOTK-UHFFFAOYSA-N | |
2-bromopyridine-3-carboxylic acid | |
233262 | |
≥98.0% (GC) | |
2-Bromonicotinic Acid |
254°C | |
MFCD01646068 | |
2-bromonicotinic acid, 2-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-bromo, 2-bromo-nicotinic acid, 2-bromo-3-carboxypyridine, 3-bromopyridine-3-carboxylic acid, bromo-nicotinic acid, pubchem5071, acmc-1aj4p, ksc223s8f | |
C1=CC(=C(N=C1)Br)C(=O)O | |
202.007 | |
202.01 | |
Crystalline Powder |
Safety and Handling
TSCA : No