missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromopentane, 90%, tech., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107235000

 View more versions of this product

Catalog No. AC107235000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromopentane
107-81-3
100.0
1.2200g/mL
20°C
12% max. (3-Bromopentane) Typical (GC)
C5H11Br
CH3CH2CH2CH(Br)CH3
500 mL
1.22
Solubility in water: practically insoluble. Other solubilities: miscible with organic solvents
CCCC(C)Br
151.05
151.05
Technical
tech., 90%
88.0
-95.5°C
116.0°C to 117.0°C
Authentic
Glass bottle
1.4393 to 1.4413
MFCD00000160
01, 131
pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
LGAJYTCRJPCZRJ-UHFFFAOYSA-N
2-bromopentane
7890
90%
Liquid

Chemical Identifiers

107-81-3
151.05
LGAJYTCRJPCZRJ-UHFFFAOYSA-N
7890
CCCC(C)Br
C5H11Br
MFCD00000160
pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
2-bromopentane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-521-3

RTECSNumber : RZ9800000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only