missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromophenol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 157360500

 View more versions of this product

Catalog No. AC157360500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromophenol
95-56-7
100.0
Colorless to Yellow
195°C
Authentic
Glass bottle
1.5880 to 1.59
MFCD00002146
06, 197
15, 1439
Solubility in water: soluble in water. Other solubilities: miscible with chloroform and ether
C1=CC=C(C(=C1)O)Br
173.01
173.01
Liquid
98%
97.5
5°C
1.4900g/mL
42°C
97.5% min. (GC)
C6H5BrO
BrC6H4OH
50 mL
1.49
o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
VADKRMSMGWJZCF-UHFFFAOYSA-N
2-bromophenol
7244
98%

Chemical Identifiers

95-56-7
173.01
VADKRMSMGWJZCF-UHFFFAOYSA-N
7244
C1=CC=C(C(=C1)O)Br
C6H5BrO
MFCD00002146
o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
2-bromophenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-432-7

RTECSNumber : SJ7875000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only