missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenol, 98%

CAS: 95-56-7 | C6H5BrO | 173.009 g/mol

$218.90 - $3128.12

Chemical Identifiers

CAS 95-56-7
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.009
MDL Number MFCD00002146
InChI Key VADKRMSMGWJZCF-UHFFFAOYSA-N
Synonym o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
PubChem CID 7244
IUPAC Name 2-bromophenol
SMILES C1=CC=C(C(=C1)O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1411214
View Documents
Thermo Scientific Chemicals
A1411214
25 g
Each for $218.90
Only null left
 
AAA1411222
View Documents
Thermo Scientific Chemicals
A1411222
100 g
Each for $718.48
Only null left
 
AAA1411236
View Documents
Thermo Scientific Chemicals
A1411236
500 g
Each for $3,128.12
Only null left
 
Description

Description

2-Bromophenol used as a disinfection byproduct found in chlorinated pool water. Used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromophenol used as a disinfection byproduct found in chlorinated pool water. Used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

Solubility
Soluble in chloroform and ether. Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed containers.
Specifications

Chemical Identifiers

95-56-7
173.009
VADKRMSMGWJZCF-UHFFFAOYSA-N
7244
C1=CC=C(C(=C1)O)Br
C6H5BrO
MFCD00002146
o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
2-bromophenol

Specifications

95-56-7
1.636
92°C (192°F)
C6H5BrO
MFCD00002146
UN3082
Light sensitive
o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
VADKRMSMGWJZCF-UHFFFAOYSA-N
2-bromophenol
7244
98%
5°C
194°C to 195°C
Phenol-like
1.589
25 g
1905115
14,1428
Soluble in chloroform and ether. Slightly soluble in water.
C1=CC=C(C(=C1)O)Br
173.009
173.01
2-Bromophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 202-432-7

RTECSNumber : SJ7875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only