missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetic acid, 98+%

CAS: 18698-97-0 | C8H7BrO2 | 215.046 g/mol

$70.45 - $579.01

Chemical Identifiers

CAS 18698-97-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00004314
InChI Key DWXSYDKEWORWBT-UHFFFAOYSA-N
Synonym 2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg
PubChem CID 87754
IUPAC Name 2-(2-bromophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1690006
View Documents
Thermo Scientific Chemicals
A1690006
5 g
Each for $70.45
Only null left
 
AAA1690014
View Documents
Thermo Scientific Chemicals
A1690014
25 g
Each for $213.60
Only null left
 
AAA1690022
View Documents
Thermo Scientific Chemicals
A1690022
100 g
Each for $579.01
Only null left
 
Description

Description

Employed as a pharmaceutical intermediate. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.

Solubility
Very faint turbidity in methanol.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications

Chemical Identifiers

18698-97-0
215.046
DWXSYDKEWORWBT-UHFFFAOYSA-N
87754
C1=CC=C(C(=C1)CC(=O)O)Br
C8H7BrO2
MFCD00004314
2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg
2-(2-bromophenyl)acetic acid

Specifications

18698-97-0
C8H7BrO2
5 g
2-bromophenylacetic acid, 2-2-bromophenyl acetic acid, o-bromophenylacetic acid, 2-bromophenyl acetic acid, benzeneacetic acid, 2-bromo, 2-bromophenylaceticacid, 2-bromo-phenyl-acetic acid, pubchem7214, acmc-1btqg
DWXSYDKEWORWBT-UHFFFAOYSA-N
2-(2-bromophenyl)acetic acid
87754
≥98%
105°C to 106°C
MFCD00004314
2250655
Very faint turbidity in methanol.
C1=CC=C(C(=C1)CC(=O)O)Br
215.046
215.05
2-Bromophenylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 242-509-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only