missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromophenylboronic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 379990250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromophenylboronic acid | |
109.0°C to 114.0°C | |
Authentic | |
Glass bottle | |
BrC6H4B(OH)2 | |
25 g | |
PLVCYMZAEQRYHJ-UHFFFAOYSA-N | |
(2-bromophenyl)boronic acid | |
2773294 | |
97% |
244205-40-1 | |
White | |
96% min. (HPLC) | |
C6H6BBrO2 | |
MFCD01114672 | |
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid | |
OB(O)C1=CC=CC=C1Br | |
200.83 | |
200.83 | |
Powder |
Chemical Identifiers
244205-40-1 | |
200.83 | |
PLVCYMZAEQRYHJ-UHFFFAOYSA-N | |
2773294 | |
OB(O)C1=CC=CC=C1Br |
C6H6BBrO2 | |
MFCD01114672 | |
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid | |
(2-bromophenyl)boronic acid |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only