Learn More
2-Bromopropene, 99%, stab.
CAS: 557-93-7 | C3H5Br | 120.977 g/mol
Supplier: Thermo Scientific Chemicals L1422922
Description
2-Bromopropene is used as a solvent and as an intermediate for organic synthesis. It is also used as a fumigant. Further, it reacts with nonanal to prepare 2-methyl-undec-1-en-3-ol using chromium(II) chloride as a reagent. It is utilized as a refrigerant used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromopropene is used as a solvent and as an intermediate for organic synthesis. It is also used as a fumigant. Further, it reacts with nonanal to prepare 2-methyl-undec-1-en-3-ol using chromium(II) chloride as a reagent. It is utilized as a refrigerant used in organic synthesis.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications
| 2-Bromopropene | |
| 557-93-7 | |
| 1.362 | |
| 4°C (39°F) | |
| C3H5Br | |
| MFCD00000140 | |
| UN1993 | |
| Light sensitive | |
| Immiscible with water. | |
| CC(=C)Br | |
| 120.977 | |
| 120.98 |
| stabilized | |
| -87°C | |
| 47°C to 49°C | |
| Pungent | |
| 1.443 | |
| 100 g | |
| 1731926 | |
| 2-bromopropene, isopropenyl bromide, 1-propene, 2-bromo, 2-bromo-1-propene, 2-bromopropylene, propene, 2-bromo, isopropylene bromide, alpha-methylvinyl bromide, .alpha.-methylvinyl bromide, 2-bromo-propene | |
| PHMRPWPDDRGGGF-UHFFFAOYSA-N | |
| 2-bromoprop-1-ene | |
| 11202 | |
| 99% |
Chemical Identifiers
| 557-93-7 | |
| 120.977 | |
| PHMRPWPDDRGGGF-UHFFFAOYSA-N | |
| 11202 | |
| CC(=C)Br |
| C3H5Br | |
| MFCD00000140 | |
| 2-bromopropene, isopropenyl bromide, 1-propene, 2-bromo, 2-bromo-1-propene, 2-bromopropylene, propene, 2-bromo, isopropylene bromide, alpha-methylvinyl bromide, .alpha.-methylvinyl bromide, 2-bromo-propene | |
| 2-bromoprop-1-ene |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 209-185-4
RTECSNumber : UC7085000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only