missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromopyridine-4-boronic acid, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 458532-94-0 |
---|---|
Molecular Formula | C5H5BBrNO2 |
Molecular Weight (g/mol) | 201.81 |
MDL Number | MFCD06798244 |
InChI Key | DFIKFQMGZNTFLI-UHFFFAOYSA-N |
Synonym | 2-bromopyridine-4-boronic acid, 2-bromopyridin-4-yl boronic acid, boronic acid, 2-bromo-4-pyridinyl-9ci, 2-bromo-4-pyridyl boronic acid, 2-bromo-4-pyridineboronic acid, boronicacid, 2-bromo-4-pyridinyl-9ci, acmc-209k4o, ksc234s1f, 2-bromo-4-pyridinyl boronic acid, 2-bromanylpyridin-4-yl boronic acid |
PubChem CID | 12060162 |
IUPAC Name | (2-bromopyridin-4-yl)boronic acid |
SMILES | OB(O)C1=CC(Br)=NC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC435920010
|
Thermo Scientific Chemicals
435920010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
458532-94-0 | |
201.81 | |
DFIKFQMGZNTFLI-UHFFFAOYSA-N | |
12060162 | |
OB(O)C1=CC(Br)=NC=C1 |
C5H5BBrNO2 | |
MFCD06798244 | |
2-bromopyridine-4-boronic acid, 2-bromopyridin-4-yl boronic acid, boronic acid, 2-bromo-4-pyridinyl-9ci, 2-bromo-4-pyridyl boronic acid, 2-bromo-4-pyridineboronic acid, boronicacid, 2-bromo-4-pyridinyl-9ci, acmc-209k4o, ksc234s1f, 2-bromo-4-pyridinyl boronic acid, 2-bromanylpyridin-4-yl boronic acid | |
(2-bromopyridin-4-yl)boronic acid |
Specifications
458532-94-0 | |
100.0 | |
94% min. (HPLC) | |
C5H5BBrNO2 | |
1g | |
DFIKFQMGZNTFLI-UHFFFAOYSA-N | |
(2-bromopyridin-4-yl)boronic acid | |
12060162 | |
95% |
94.0 | |
Authentic | |
Glass Bottle | |
MFCD06798244 | |
2-bromopyridine-4-boronic acid, 2-bromopyridin-4-yl boronic acid, boronic acid, 2-bromo-4-pyridinyl-9ci, 2-bromo-4-pyridyl boronic acid, 2-bromo-4-pyridineboronic acid, boronicacid, 2-bromo-4-pyridinyl-9ci, acmc-209k4o, ksc234s1f, 2-bromo-4-pyridinyl boronic acid, 2-bromanylpyridin-4-yl boronic acid | |
OB(O)C1=CC(Br)=NC=C1 | |
201.81 | |
201.82 | |
2-Bromopyridine-4-boronic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only