missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoquinoxaline 98.0+%, TCI America™

$59.65 - $231.14

Chemical Identifiers

CAS 36856-91-4
Molecular Formula C8H5BrN2
Molecular Weight (g/mol) 209.046
MDL Number MFCD07368206
InChI Key XVKLMYQMVPHUPN-UHFFFAOYSA-N
Synonym quinoxaline, 2-bromo, bromoquinoxaline, 2-bromoquinoxaline, ksc226a0j
PubChem CID 582225
IUPAC Name 2-bromoquinoxaline
SMILES C1=CC=C2C(=C1)N=CC(=N2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4632200MG
View Documents
TCI America
B4632200MG
200 mg
Each for $59.65
Only null left
 
B46321G
View Documents
TCI America
B46321G
1 g
Each for $231.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

36856-91-4
209.046
XVKLMYQMVPHUPN-UHFFFAOYSA-N
582225
C1=CC=C2C(=C1)N=CC(=N2)Br
C8H5BrN2
MFCD07368206
quinoxaline, 2-bromo, bromoquinoxaline, 2-bromoquinoxaline, ksc226a0j
2-bromoquinoxaline

Specifications

36856-91-4
C8H5BrN2
200 mg
XVKLMYQMVPHUPN-UHFFFAOYSA-N
2-bromoquinoxaline
582225
≥98.0% (GC)
2-Bromoquinoxaline
57°C
MFCD07368206
quinoxaline, 2-bromo, bromoquinoxaline, 2-bromoquinoxaline, ksc226a0j
C1=CC=C2C(=C1)N=CC(=N2)Br
209.046
209.05
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents