missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromothiazole-4-carboxaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 5198-80-1 |
---|---|
Molecular Formula | C4H2BrNOS |
Molecular Weight (g/mol) | 192.03 |
MDL Number | MFCD04115726 |
InChI Key | JDUXMFGFGCJNGO-UHFFFAOYSA-N |
Synonym | 2-bromothiazole-4-carbaldehyde, 2-bromo-4-formylthiazole, 2-bromo-4-fomylthiazole, 2-bromothiazole-4-carboxaldehyde, 4-thiazolecarboxaldehyde, 2-bromo, 2-bromo-1,3-thiazole-4-carboxaldehyde, 2-bromo-4-formyl-1,3-thiazole, pubchem8915, acmc-209kx4, 2-bromo-4-thiazole carboxaldehyde |
PubChem CID | 17750433 |
IUPAC Name | 2-bromo-1,3-thiazole-4-carbaldehyde |
SMILES | BrC1=CSC(C=O)=N1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC429540010
|
Thermo Scientific Chemicals
429540010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC429540050
|
Thermo Scientific Chemicals
429540050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5198-80-1 | |
192.03 | |
JDUXMFGFGCJNGO-UHFFFAOYSA-N | |
17750433 | |
BrC1=CSC(C=O)=N1 |
C4H2BrNOS | |
MFCD04115726 | |
2-bromothiazole-4-carbaldehyde, 2-bromo-4-formylthiazole, 2-bromo-4-fomylthiazole, 2-bromothiazole-4-carboxaldehyde, 4-thiazolecarboxaldehyde, 2-bromo, 2-bromo-1,3-thiazole-4-carboxaldehyde, 2-bromo-4-formyl-1,3-thiazole, pubchem8915, acmc-209kx4, 2-bromo-4-thiazole carboxaldehyde | |
2-bromo-1,3-thiazole-4-carbaldehyde |
Specifications
5198-80-1 | |
100.0 | |
96% min. (HPLC) | |
C4H2BrNOS | |
1g | |
JDUXMFGFGCJNGO-UHFFFAOYSA-N | |
2-bromo-1,3-thiazole-4-carbaldehyde | |
17750433 | |
97% |
96.0 | |
Authentic | |
Glass bottle | |
MFCD04115726 | |
2-bromothiazole-4-carbaldehyde, 2-bromo-4-formylthiazole, 2-bromo-4-fomylthiazole, 2-bromothiazole-4-carboxaldehyde, 4-thiazolecarboxaldehyde, 2-bromo, 2-bromo-1,3-thiazole-4-carboxaldehyde, 2-bromo-4-formyl-1,3-thiazole, pubchem8915, acmc-209kx4, 2-bromo-4-thiazole carboxaldehyde | |
BrC1=CSC(C=O)=N1 | |
192.03 | |
192.03 | |
2-Bromothiazole-4-carboxaldehyde |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
RUO â Research Use Only