missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole, 99%

CAS: 3034-53-5 | C3H2BrNS | 164.02 g/mol

$80.49 - $268.33

Chemical Identifiers

CAS 3034-53-5
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD00005316
InChI Key RXNZFHIEDZEUQM-UHFFFAOYSA-N
Synonym 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
PubChem CID 76430
IUPAC Name 2-bromo-1,3-thiazole
SMILES C1=CSC(=N1)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1483809
View Documents
Thermo Scientific Chemicals
A1483809
10 g
Each for $80.49
Only null left
 
AAA1483814
View Documents
Thermo Scientific Chemicals
A1483814
25 g
Each for $160.12
Only null left
 
AAA1483818
View Documents
Thermo Scientific Chemicals
A1483818
50 g
Each for $268.33
Only null left
 
Description

Description

2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Hydrogen bromide gas, nitrogen oxides (NOx), Sulphur oxides.
Specifications

Chemical Identifiers

3034-53-5
164.02
RXNZFHIEDZEUQM-UHFFFAOYSA-N
76430
C1=CSC(=N1)Br
C3H2BrNS
MFCD00005316
2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
2-bromo-1,3-thiazole

Specifications

3034-53-5
171°C
C3H2BrNS
MFCD00005316
105724
It is insoluble in water.
C1=CSC(=N1)Br
164.02
164.02
2-Bromothiazole
1.836
63°C (145°F)
1.593
10 g
2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
RXNZFHIEDZEUQM-UHFFFAOYSA-N
2-bromo-1,3-thiazole
76430
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 221-229-4

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only