missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromothiobenzamide, 97%, Thermo Scientific™
$60.58 - $60.58
Chemical Identifiers
CAS | 30216-44-5 |
---|---|
Molecular Formula | C7H6BrNS |
Molecular Weight (g/mol) | 216.096 |
MDL Number | MFCD04973314 |
InChI Key | GHCQWSAFBXLYSO-UHFFFAOYSA-N |
Synonym | 2-bromothiobenzamide, 2-bromobenzothioamide, thiobenzamide, 2-bromo, 2-bromobenzene-1-carbothioamide, benzenecarbothioamide,2-bromo, amino 2-bromophenyl methane-1-thione, 2-bromo-thiobenzamide, acmc-1agnp |
PubChem CID | 2734818 |
IUPAC Name | 2-bromobenzenecarbothioamide |
SMILES | C1=CC=C(C(=C1)C(=S)N)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH55238MD
|
Thermo Scientific Chemicals
H55238MD |
250 mg |
Each for $60.58
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
30216-44-5 | |
216.096 | |
GHCQWSAFBXLYSO-UHFFFAOYSA-N | |
2734818 | |
C1=CC=C(C(=C1)C(=S)N)Br |
C7H6BrNS | |
MFCD04973314 | |
2-bromothiobenzamide, 2-bromobenzothioamide, thiobenzamide, 2-bromo, 2-bromobenzene-1-carbothioamide, benzenecarbothioamide,2-bromo, amino 2-bromophenyl methane-1-thione, 2-bromo-thiobenzamide, acmc-1agnp | |
2-bromobenzenecarbothioamide |
Specifications
30216-44-5 | |
Brown to White | |
MFCD04973314 | |
2-bromothiobenzamide, 2-bromobenzothioamide, thiobenzamide, 2-bromo, 2-bromobenzene-1-carbothioamide, benzenecarbothioamide,2-bromo, amino 2-bromophenyl methane-1-thione, 2-bromo-thiobenzamide, acmc-1agnp | |
C1=CC=C(C(=C1)C(=S)N)Br | |
216.096 | |
216.1 | |
Solid |
82°C to 86°C | |
C7H6BrNS | |
250 mg | |
GHCQWSAFBXLYSO-UHFFFAOYSA-N | |
2-bromobenzenecarbothioamide | |
2734818 | |
97% | |
2-Bromothiobenzamide |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures