missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Butanol 99.0+%, TCI America™
$39.51 - $53.32
Chemical Identifiers
CAS | 78-92-2 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00004569 |
InChI Key | BTANRVKWQNVYAZ-UHFFFAOYNA-N |
Synonym | 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol |
PubChem CID | 6568 |
ChEBI | CHEBI:35687 |
IUPAC Name | butan-2-ol |
SMILES | CCC(C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B070525ML
|
TCI America
B070525ML |
25 mL |
Each for $39.51
|
|
|||||
B0705500ML
|
TCI America
B0705500ML |
500 mL |
Each for $53.32
|
|
|||||
Chemical Identifiers
78-92-2 | |
74.12 | |
BTANRVKWQNVYAZ-UHFFFAOYNA-N | |
6568 | |
butan-2-ol |
C4H10O | |
MFCD00004569 | |
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol | |
CHEBI:35687 | |
CCC(C)O |
Specifications
78-92-2 | |
Colorless | |
C4H10O | |
25 mL | |
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol | |
CCC(C)O | |
74.12 | |
CHEBI:35687 | |
≥99.0% (GC) | |
2-Butanol |
-115°C | |
100°C | |
MFCD00004569 | |
1120 | |
BTANRVKWQNVYAZ-UHFFFAOYNA-N | |
butan-2-ol | |
6568 | |
74.12 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-3049
RTECSNumber : EO1750000
TSCA : Yes