Learn More
2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 154330025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Butoxyethanol | |
98.5 | |
-70.0°C | |
171.0°C | |
Authentic | |
Glass Bottle | |
1.4180 to 1.4200 | |
MFCD00002884 | |
01, II, 519 | |
0.901 | |
butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb | |
POAOYUHQDCAZBD-UHFFFAOYSA-N | |
2-butoxyethanol | |
8133 | |
CHEBI:63921 | |
99% | |
Liquid |
111-76-2 | |
100.0 | |
0.9010g/mL | |
62°C | |
98.5% min. (GC) | |
C6H14O2 | |
CH3(CH2)3OCH2CH2OH | |
2.5 L | |
0.02% max. | |
15, 1560 | |
Solubility in water: soluble. Other solubilities: soluble in most organic solvents | |
CCCCOCCO | |
118.18 | |
5.31 mPa.s (20°C) | |
118.18 | |
Extra Pure |
Chemical Identifiers
111-76-2 | |
118.18 | |
POAOYUHQDCAZBD-UHFFFAOYSA-N | |
8133 | |
CCCCOCCO |
C6H14O2 | |
MFCD00002884 | |
butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb | |
CHEBI:63921 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Warning
EINECSNumber : 203-905-
RUO – Research Use Only