missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 154330100

 View more versions of this product

Catalog No. AC154330100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Butoxyethanol
98.5
-70.0°C
171.0°C
Authentic
Plastic drum
1.4180 to 1.4200
MFCD00002884
01, II, 519
0.901
butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
POAOYUHQDCAZBD-UHFFFAOYSA-N
2-butoxyethanol
8133
CHEBI:63921
99%
Liquid
111-76-2
100.0
0.9010g/mL
62°C
98.5% min. (GC)
C6H14O2
CH3(CH2)3OCH2CH2OH
10 L
0.02% max.
15, 1560
Solubility in water: soluble. Other solubilities: soluble in most organic solvents
CCCCOCCO
118.18
5.31 mPa.s (20°C)
118.18
Extra Pure

Chemical Identifiers

111-76-2
118.18
POAOYUHQDCAZBD-UHFFFAOYSA-N
8133
CCCCOCCO
C6H14O2
MFCD00002884
butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
CHEBI:63921
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Warning

EINECSNumber : 203-905-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only