Learn More
2-Butoxyethyl acetate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 403410100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2-Butoxyethyl acetate | |
| 112-07-2 | |
| 100.0 | |
| 193.5°C | |
| Authentic | |
| Plastic drum | |
| 1.4120 to 1.4140 | |
| MFCD00009457 | |
| 0.9385 | |
| Solubility in water: soluble | |
| CCCCOCCOC(C)=O | |
| 160.21 | |
| 160.21 |
| 98% | |
| 97.5 | |
| 0.9385g/mL | |
| 76°C | |
| 97.5% min. (GC) | |
| C8H16O3 | |
| CH3CO2CH2CH2O(CH2)3CH3 | |
| 10L | |
| butoxyethyl acetate, butyl glycol acetate, butylglycol acetate, ethylene glycol monobutyl ether acetate, 2-butoxyethanol acetate, butyl cellosolve acetate, ektasolve eb acetate, ethanol, 2-butoxy-, acetate, butylcelosolvacetat, 1-acetoxy-2-butoxyethane | |
| NQBXSWAWVZHKBZ-UHFFFAOYSA-N | |
| 2-butoxyethyl acetate | |
| 8160 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
EINECSNumber : 203-933-3
RTECSNumber : KJ8925000
TSCA : TSCA
RUO â Research Use Only